missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Human Integrin alpha 4 beta 1 Protein
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
Beschreibung
The Recombinant Human Integrin alpha 4 beta 1 Protein is derived from CHO. The Recombinant Human Integrin alpha 4 beta 1 Protein has been validated for the following applications: Bioactivity.
Spezifikation
Spezifikation
| Formulation | Lyophilized from a 0.2μm filtered solution in PBS. |
| Gene ID (Entrez) | 3676 |
| Molecular Weight (g/mol) | 108.8 kDa ( alpha4) & 82.3 kDa (beta1) |
| Name | Integrin alpha 4 beta 1 |
| Quantity | 50 μg |
| Source | Chinese Hamster Ovary cell line,CHO-derived human Integrin alpha 4 beta 1 protein, (N-terminus) Human Integrin alpha 4 (Tyr34-Gln970 (Arg591Leu & Arg878Gln)) Accession &Num; P13612 Acidic Tail 6-His tag Human Integrin beta 1 (Gln21-Asp728) Accession &Num; P05556 Basic Tail (C-terminus) |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. |
| Gene Alias | CD49d, IA4 |
| Conjugate | Unconjugated |
| Recombinant | Recombinant |
| Mehr anzeigen |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur