missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Human Integrin alpha M beta 2 Protein
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
Description
The Recombinant Human Integrin alpha M beta 2 Protein is derived from CHO. The Recombinant Human Integrin alpha M beta 2 Protein has been validated for the following applications: Bioactivity.
Specifications
Specifications
| Formulation | Lyophilized from a 0.2 μm filtered solution in Tris, NaCl and MgCl 2 . |
| Gene ID (Entrez) | 3684 |
| Molecular Weight (g/mol) | 124.7 kDa (Integrin alphaM), 79 kDa (Integrin beta2) |
| Name | Integrin alpha M beta 2 |
| Quantity | 50 μg |
| Source | Chinese Hamster Ovary cell line,CHO-derived human Integrin alpha M beta 2 protein, (N-terminus) Human Integrin alpha M (Phe17-Asn1105) Accession &Num; NP_001139280 acidic tail Human Integrin beta 2 (Gln23-Asn700) Accession &Num; P05107 basic tail (C-terminus) |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. |
| Conjugate | Unconjugated |
| Recombinant | Recombinant |
| Purity or Quality Grade | 95%, by SDS-PAGE under reducing conditions and visualized by silver stain. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction