missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Human Integrin alpha V beta 3 Fc Protein, CF

Product Code. 30306763
Change view
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30306763 50 μg 50µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30306763 Supplier R&D Systems™ Supplier No. 11648AV050
 Restricted Item
Estimated Shipment: 03-12-2026
Aggiungi al carrello
Aggiungi al carrello

missing translation for 'validMainframeMsg'

Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso

Fc Chimera

Measured by its binding ability in a functional ELISA. Recombinant Human Integrin alpha V beta 3 Fc Chimera (Catalog # 11648-AV) binds to Recombinant Human Vitronectin (Catalog # 2308-VN ) with an ED50 of 60.0-600 ng/mL.
TRUSTED_SUSTAINABILITY

Specifica

For Use With (Application) Bioactivity
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose.
Gene ID (Entrez) 3685
Molecular Weight (g/mol) MolecularWeight-observed: >190 kDa, under non-reducing conditions., MolecularWeight-theroretical: 133 kDa (Integrin alpha V) & 103 kDa (Integrin beta 3)
Quantity 50 μg
Source Human embryonic kidney cell, HEK293-derived human Integrin alpha V beta 3 protein Human ITGAV (Phe31-Val992) (N-terminus) Accession # AAA36808.1 IEGR Human IgG1 (Glu99-Lys330) (with modifications) Human ITGB3 (Gly27-Asp718) (N-terminus) Accession # P05106.2 HPIEGR Human IgG1 (Glu99-Lys330) (with modifications) (C-terminus)
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 3 months, -20 to -70 °C under sterile conditions after reconstitution.
Endotoxin Concentration <0.10 EU per 1 μg of the protein by the LAL method.
Gene Alias antigen identified by monoclonal L230, CD51, CD51 antigen, integrin alpha-V, integrin, alpha V (vitronectin receptor, alpha polypeptide, antigen CD51), MSK8, Vitronectin receptor subunit alpha, VNRADKFZp686A08142
Conjugate Unconjugated
Reconstitution Reconstitute at 250 μg/mL in water.
Product Line R&D Systems
Protein Integrin alpha V beta 3
Vedi altri risultati Mostra meno risultati
Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.