missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Interferon epsilon Protein
A DNA sequence encoding the Homo sapiens (Human) Interferon epsilon, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7803-ec-1mg
Additional Details : Weight : 0.01000kg
Specifications
NP_795372.1 | |
Interferon epsilon Protein | |
Research Use Only | |
Ifne | |
Recombinant Protein | |
E. coli | |
LDLKLIIFQQ RQVNQESLKL LNKLQTLSIQ QCLPHRKNFL LPQKSLSPQQ YQKGHTLAIL HEMLQQIFSL FRANISLDGW EENHTEKFLI QLHQQLEYLE ALMGLEAEKL SGTLGSDNLR LQVKMYFRRI HDYLENQDYS TCAWAIVQVE ISRCLFFVFS LTEKLSKQGR PLNDMKQELT TEFRSPR | |
Greater than 80% as determined by SDS-PAGE. |
338376 | |
1 mg | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |