missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Intraflagellar transport protein 22 homolog Protein

A DNA sequence encoding the Homo sapiens (Human) Intraflagellar transport protein 22 homolog, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8067-ec-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16042471

  • 505.00 € / 100µg
Estimated Shipment: 27-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_001124292.1
Intraflagellar transport protein 22 homolog Protein
Research Use Only
RABL5
Recombinant Protein
E. coli
MLKAKILFVG PCESGKTVLA NFLTESSDIT EYSPTQGVRI LEFENPHVTS NNKGTGCEFE LWDCGGDAKF ESCWPALMKD AHGVVIVFNA DIPSHRKEME MWYSCFVQQP SLQDTQCMLI AHHKPGSGDD KGSLSLSPPL NKLKLVHSNL EDDPEEIRME FIKYLKSIIN SMSESRDREE MSIMT
Greater than 90% as determined by SDS-PAGE.
64792
100 μg
Not determined.
Full Length
Human
His-SUMO
Tris-based buffer, 50% glycerol
Suggestions de produits

Suggestions de produits

Vidéos
FDS
Documents

Documents

Certificats
Promotions

Promotions