missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Involucrin Protein

Product Code. p-7150316
Click to view available options
Quantity:
10 μg
2 μg
5 μg
Unit Size:
10µg
2µg
5µg
This item is not returnable. View return policy

Product Code. 15920469

Brand: enQuireBio™ QP124532ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

A cDNA sequence encoding the Involucrin was constructed and used to recombinantly synthesize the protein.

Specifications

Gene ID (Entrez) 3713
Name Involucrin Protein
Quantity 2 μg
Regulatory Status Research Use Only
Endotoxin Concentration Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Gene Symbol IVL
Product Type Recombinant Protein
Cross Reactivity Human
Species E. coli
Protein Tag GST
Sequence MSQQHTLPVT LSPALSQELL KTVPPPVNTH QEQMKQPTPL PPPCQKVPVE LPVEVPSKQE EKHMTAVKGL PEQECEQQQK EPQEQELQQQ HWEQHEEYQK AENPEQQLKQ EKTQRDQQLN KQLEEEKKLL DQQLDQELVK RDEQLGMKKE QLLELPEQQE GHLKHLEQQE GQLKHPEQQE GQLELPEQQE GQLELPEQQE GQLELPEQQE GQLELPEQQE GQLELPEQQE GQLELPQQQE GQLELSEQQE GQLELSEQQE GQLKHLEHQE GQLEVPEEQM GQLKYLEQQE GQLKHLDQQE KQPELPEQQM GQLKHLEQQE GQPKHLEQQE GQLEQLEEQE GQLKHLEQQE GQLEHLEHQE GQLGLPEQQV LQLKQLEKQQ GQPKHLEEEE GQLKHLVQQE GQLKHLVQQE GQLEQQERQV EHLEQQVGQL KHLEEQEGQL KHLEQQQGQL EVPEQQVGQP KNLEQEEKQL ELPEQQEGQV KHLEKQEAQL ELPEQQVGQP KHLEQQEKHL EHPEQQDGQL KHLEQQEGQL KDLEQQKGQL EQPVFAPAPG QVQDIQPALP TKGEVLLPVE HQQQKQEVQW PPKHK
Buffer Involucrin at 100microg/ml in 50mM Tris-HCl, pH 7.5, 10mM L-glutathione (reduced).
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.