missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human IPP-POZ Protein
A cDNA sequence encoding the IPP-POZ was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP12455-50ug
Additional Details : Weight : 0.01000kg
Specifications
IPP-POZ Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MANEDCPKAA DSPFSSDKHA QLILAQINKM RNGQHFCDVQ LQVGQESFKA HRLVLAASSPYFAALFTGGM KESSKDVVPI LGIEAGIFQI LLDFIYTGIV NIGVNNVQEL IIAADMLQLTEVVHLCCEFL KGQIDPLNCI GIFQFSEQIA CHDLLEF | |
Greater than 95.0% as determined by(a) Analysis by RP-HPLC. (b) Analysis by SDS-PAGE. |
50 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
Untagged | |
The protein (1 mg/ml) containing 10mM HEPES (pH 7.4) and 25mM NaCl. |