missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human ITLN1 / Omentin Protein
Recombinant Protein
107.00 € - 7105.00 €
Specifications
Accession Number | NP_060095.2 |
---|---|
Gene ID (Entrez) | 55600 |
Name | ITLN1 / Omentin Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Less than or equal to 5.00 EUs/ug as determined by the LAL method. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15985789
|
enQuireBio™
QP5262-2UG |
2 μg |
107.00 €
2µg |
Estimated Shipment: 21-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15965789
|
enQuireBio™
QP5262-10UG |
10 μg |
252.00 €
10µg |
Estimated Shipment: 21-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15955789
|
enQuireBio™
QP5262-100UG |
100 μg |
940.00 €
100µg |
Estimated Shipment: 21-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15995789
|
enQuireBio™
QP5262-500UG |
500 μg |
4325.00 €
500µg |
Estimated Shipment: 21-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15975789
|
enQuireBio™
QP5262-1MG |
1 mg |
7105.00 €
1mg |
Estimated Shipment: 21-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
NP_060095.2 | |
ITLN1 / Omentin Protein | |
Less than or equal to 5.00 EUs/ug as determined by the LAL method. | |
Recombinant Protein | |
E. coli | |
MNQLSFLLFL IATTRGWSTD EANTYFKEWT CSSSPSLPRS CKEIKDECPS AFDGLYFLRT ENGVIYQTFC DMTSGGGGWT LVASVHENDM RGKCTVGDRW SSQQGSKADY PEGDGNWANY NTFGSAEAAT SDDYKNPGYY DIQAKDLGIW HVPNKSPMQH WRNSSLLRYR TDTGFLQTLG HNLFGIYQKY PVKYGEGKCW TDNGPVIPVV YDFGDAQKTA SYYSPYGQRE FTAGFVQFRV FNNERAANAL CAGMRVTGCN TEHHCIGGGG YFPEASPQQC GDFSGFDWSG YGTHVGYSSS REITEAAVLL FYR | |
Greater than 90 as determined by SDS-PAGE. |
55600 | |
Research Use Only | |
ITLN1 | |
Human | |
Untagged | |
10 mM sodium phosphate and 5:1 mannitol to protein, pH 7.5 |