missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human ITPA Protein
A cDNA sequence encoding the ITPA was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP12464-5ug
Additional Details : Weight : 0.01000kg
Specifications
ITPA Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MMAASLVGKK IVFVTGNAKK LEEVVQILGD KFPCTLVAQK IDLPEYQGEP DEISIQKCQE AVRQVQGPVL VEDTCLCFNA LGGLPGPYIK WFLEKLKPEG LHQLLAGFED KSAYALCTFA LSTGDPSQPV RLFRGRTSGR IVAPRGCQDF GWDPCFQPDG YEQTYAEMPK AEKNAVSHRF RALLELQEYF GSLAA | |
Greater than 95.0% as determined by SDS-PAGE. |
5 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The ITPA solution (1 mg/ml) contains 20mM Tris-HCl pH 8 and 10% glycerol. |