missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human JAM3 Protein
A cDNA sequence encoding the JAM3 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP12468-2ug
Additional Details : Weight : 0.01000kg
Specifications
83700 | |
2 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMVNLKSS NRTPVVQEFE SVELSCIITD SQTSDPRIEW KKIQDEQTTY VFFDNKIQGD LAGRAEILGK TSLKIWNVTR RDSALYRCEV VARNDRKEID EIVIELTVQV KPVTPVCRVP KAVPVGKMAT LHCQESEGHP RPHYSWYRND VPLPTDSRAN PRFRNSSFHL NSETGTLVFT AVHKDDSGQY YCIASNDAGS ARCEEQEMEV YDLN | |
Greater than 90% as determined by SDS-PAGE. |
JAM3 Protein | |
Research Use Only | |
JAM3 | |
Human | |
His | |
The JAM3 solution contains 20mM Tris-HCl buffer (pH 8.0), 0.2M NaCl, 2mM EDTA, 5mM DTT and 50% glycerol. |