missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human JSRP1 Protein

A cDNA sequence encoding the JSRP1 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP12473-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15990529

  • 1740.00 € / 100µg
Estimated Shipment: 20-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

JSRP1 Protein
Research Use Only
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MGSMSMTTRA WEELDGGLGS CQALEDHSAL AETQEDRASA TPRLADSGSV PHDSQVAEGP SVDTRPKKME KEPAARGTPG TGKERLKAGA SPRSVPARKK AQTAPPLQPP PPPPALSEEL PWGDLSLNKC LVLASLVALL GSAFQLCRDA VPGEAALQAR VPEPWVPPSS APREPSSPLP KFEAQAPPSA PPAPRAEAEV RPKIPGSREA AENDEEEPGE ATGEAVREDR VTLADRGPKE RPRREGKPRK EKPRKEERPK KERPRKEERP RAAREPREAL PQRWESREGG HRPWARDSRD AEPRKKQAWV SPRRPDEEQR PGSRQKLRAG KGRD
Greater than 90.0% as determined by SDS-PAGE.
100 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
The JSRP1 solution (0.25 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 0.15M NaCl and 10% glycerol.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers