missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Keratin, type I cuticular Ha8 Protein
A DNA sequence encoding the Homo sapiens (Human) Keratin, type I cuticular Ha8, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6280-ec-500ug
Additional Details : Weight : 0.01000kg
Specifications
NP_006762.3 | |
Keratin, type I cuticular Ha8 Protein | |
Research Use Only | |
KRT38 | |
Recombinant Protein | |
E. coli | |
TSSYSSSSCP LGCTMAPGAR NVSVSPIDIG CQPGAEANIA PMCLLANVAH ANRVRVGSTP LGRPSLCLPP TCHTACPLPG TCHIPGNIGI CGAYGENTLN GHEKETMQFL NDRLANYLEK VRQLEQENAE LEATLLERSK CHESTVCPDY QSYFHTIEEL QQKILCSKAE NARLIVQIDN AKLAADDFRI KLESERSLRQ LVEADKCGTQ KLLDDATLAK ADLEAQQESL KEEQLSLKSN HEQEVKILRS QLGEKLRIEL DIEPTIDLNR VLGEMRAQYE AMLETNRQDV EQWFQAQSEG ISLQDMSCSE ELQCCQSEIL ELRCTVNALE VERQAQHTLK DCLQNSLCEA EDRFGTELAQ MQSLISNVEE QLSEIRADLE RQNQEYQVLL DVKTRLENEI ATYRNLLESE DCKLPCNPCS TSPSCVTAPC APRPSCGPCT TCGPTCGAST TGSRF | |
Greater than 90% as determined by SDS-PAGE. |
8687 | |
500 μg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |