missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Keratin, type I cuticular Ha8 Protein
A DNA sequence encoding the Homo sapiens (Human) Keratin, type I cuticular Ha8, was expressed in the hosts and tags indicated.
160.00 € - 2285.00 €
Specifications
Accession Number | NP_006762.3 |
---|---|
Gene ID (Entrez) | 8687 |
Name | Keratin, type I cuticular Ha8 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16012150
|
enQuireBio™
QP6280-EC-100UG |
100 μg |
505.00 €
100µg |
Estimated Shipment: 21-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16032150
|
enQuireBio™
QP6280-EC-1MG |
1 mg |
2285.00 €
1mg |
Estimated Shipment: 21-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16022150
|
enQuireBio™
QP6280-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16042150
|
enQuireBio™
QP6280-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16052150
|
enQuireBio™
QP6280-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16062150
|
enQuireBio™
QP6280-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_006762.3 | |
Keratin, type I cuticular Ha8 Protein | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
8687 | |
Research Use Only | |
KRT38 | |
Recombinant Protein | |
E. coli | |
TSSYSSSSCP LGCTMAPGAR NVSVSPIDIG CQPGAEANIA PMCLLANVAH ANRVRVGSTP LGRPSLCLPP TCHTACPLPG TCHIPGNIGI CGAYGENTLN GHEKETMQFL NDRLANYLEK VRQLEQENAE LEATLLERSK CHESTVCPDY QSYFHTIEEL QQKILCSKAE NARLIVQIDN AKLAADDFRI KLESERSLRQ LVEADKCGTQ KLLDDATLAK ADLEAQQESL KEEQLSLKSN HEQEVKILRS QLGEKLRIEL DIEPTIDLNR VLGEMRAQYE AMLETNRQDV EQWFQAQSEG ISLQDMSCSE ELQCCQSEIL ELRCTVNALE VERQAQHTLK DCLQNSLCEA EDRFGTELAQ MQSLISNVEE QLSEIRADLE RQNQEYQVLL DVKTRLENEI ATYRNLLESE DCKLPCNPCS TSPSCVTAPC APRPSCGPCT TCGPTCGAST TGSRF | |
Greater than 90% as determined by SDS-PAGE. |