missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Human KGF/FGF-7 Protein

Product Code. 18765905
Click to view available options
Quantity:
1 mg
10 μg
100 μg
50 μg
500 μg
Unit Size:
100µg
10µg
1mg
500µg
50µg
This item is not returnable. View return policy

Product Code. 18765905

Brand: R&D Systems™ 11340KG500

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Carrier Free Recombinant Protein

Measured in a serum-free cell proliferation assay using MCF-7 human breast cancer cells. Karey, K.P. et al. (1988) Cancer Research 48:4083. The ED50 for this effect is 25.0-250 ng/mL.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P21781.1
Formulation Lyophilized from a 0.2 μm filtered solution in MOPS, Na2SO4 and EDTA with Trehalose.
Gene ID (Entrez) 2252
Molecular Weight (g/mol) MolecularWeight-observed: 22-28 kDa, under reducing conditions., MolecularWeight-theroretical: 19 kDa
Quantity 500 μg
Source Human embryonic kidney cell, HEK293-derived human KGF/FGF-7 protein Cys32-Thr194
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution.
Endotoxin Concentration <0.10 EU / 1 μg of the protein by the LAL method.
Gene Alias FGF7, FGF-7, fibroblast growth factor 7 HBGF-7, HBGF7, HBGF-7, Heparin-binding growth factor 7, keratinocyte growth factor, KGFfibroblast growth factor 7 (keratinocyte growth factor)
Conjugate Unconjugated
Reconstitution Reconstitute 10 μg size at 100 μg/mL and all the other sizes (at 250 μg/mL in PBS.)
Purity or Quality Grade >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
Protein KGF/FGF-7
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.