missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Kinesin light chain 3 Protein

A DNA sequence encoding the Homo sapiens (Human) Kinesin light chain 3, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6265-ye-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16032040

  • 880.00 € / 100µg
Estimated Shipment: 05-07-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_803136.2
Kinesin light chain 3 Protein
Research Use Only
KLC3
Recombinant Protein
Yeast
MSVQVAAPGS AGLGPERLSP EELVRQTRQV VQGLEALRAE HHGLAGHLAE ALAGQGPAAG LEMLEEKQQV VSHSLEAIEL GLGEAQVLLA LSAHVGALEA EKQRLRSQAR RLAQENVWLR EELEETQRRL RASEESVAQL EEEKRHLEFL GQLRQYDPPA ESQQSESPPR RDSLASLFPS EEEERKGPEA AGAAAAQQGG YEIPARLRTL HNLVIQYAGQ GRYEVAVPLC RQALEDLERS SGHCHPDVAT MLNILALVYR DQNKYKEATD LLHDALQIRE QTLGPEHPAV AATLNNLAVL YGKRGRYREA EPLCQRALEI REKVLGADHP DVAKQLNNLA LLCQNQGKFE DVERHYARAL SIYEALGGPH DPNVAKTKNN LASAYLKQNK YQQAEELYKE ILHKEDLPAP LGAPNTGTAG DAEQALRRSS SLSKIRESIR RGSEKLVSRL RGEAAAGAAG MKRAMSLNTL NVDAPRAPGT QFPSWHLDKA PRTLSASTQD LSPH
Greater than 90% as determined by SDS-PAGE.
147700
100 μg
Not determined.
Full Length
Human
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers