missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human KIR2DL1 Protein

A cDNA sequence encoding the KIR2DL1 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP12487-5ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15930579

  • 131.00 € / 5µg
Estimated Shipment: 07-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

KIR2DL1 Protein
Research Use Only
Recombinant Protein
E. coli
MEGVHRKPSL LAHPGRLVKS EETVILQCWS DVMFEHFLLH REGMFNDTLR LIGEHHDGVS KANFSISRMT QDLAGTYRCY GSVTHSPYQV SAPSDPLDIV IIGLYEKPSL SAQLGPTVLA GENVTLSCSS RSSYDMYHLS REGEAHERRL PAGPKVNGTF QADFPLGPAT HGGTYRCFGS FHDSPYEWSK SSDPLLVSVT GN
Greater than 95.0% as determined by(a) Analysis by RP-HPLC. (b) Analysis by SDS-PAGE.
5 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
Untagged
The protein (1 mg/ml) contains 25mM Tris- HCl (pH 7.5).
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers