missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human KLK3 Protein
A cDNA sequence encoding the KLK3 was constructed and used to recombinantly synthesize the protein.
131.00 € - 3915.00 €
Specifications
Gene ID (Entrez) | 354 |
---|---|
Name | KLK3 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Gene Symbol | KLK3 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15910649
|
enQuireBio™
QP12506-HIS-5UG |
5 μg |
131.00 €
5µg |
Estimated Shipment: 21-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15900649
|
enQuireBio™
QP12506-HIS-20UG |
20 μg |
203.00 €
20µg |
Estimated Shipment: 21-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15990639
|
enQuireBio™
QP12506-HIS-1MG |
1 mg |
3915.00 €
1mg |
Estimated Shipment: 21-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
354 | |
Research Use Only | |
KLK3 | |
Human | |
His | |
KLK3 protein filtered (0.4microm) and lyophilized in 0.5 mg/ml in 0.05M phosphate buffer and 0.075M NaCl pH 7.4. |
KLK3 Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
IVGGWECEKH SQPWQVLVAS RGRAVCGGVL VHPQWVLTAA HCIRNKSVIL LGRHSLFHPE DTGQVFQVSH SFPHPLYDMS LLKNRFLRPG DDSSHDLMLL RLSEPAELTD AVKVMDLPTQ EPALGTTCYA SGWGSIEPEE FLTPKKLQCV DLHVISNDVC AQVHPQKVTK FMLCAGRWTG GKSTCSGDSG GPLVCNGVLQ GITSWGSEPC ALPERPSLYT KVVHYRKWIK DTIVANP | |
Greater than 90% as determined by SDS-PAGE. |