missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human L-lactate dehydrogenase C chain Protein

A DNA sequence encoding the Homo sapiens (Human) L-lactate dehydrogenase C chain, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6291-ec-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16042230

  • 3375.00 € / 1mg
Estimated Shipment: 03-08-2022
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_002292.1
L-lactate dehydrogenase C chain Protein
1 mg
Not determined.
Full Length
Human
His-SUMO
Tris-based buffer, 50% glycerol
3948
Greater than 90% as determined by SDS-PAGE.
Research Use Only
LDHC
Recombinant Protein
E. coli
STVKEQLIEK LIEDDENSQC KITIVGTGAV GMACAISILL KDLADELALV DVALDKLKGE MMDLQHGSLF FSTSKITSGK DYSVSANSRI VIVTAGARQQ EGETRLALVQ RNVAIMKSII PAIVHYSPDC KILVVSNPVD ILTYIVWKIS GLPVTRVIGS GCNLDSARFR YLIGEKLGVH PTSCHGWIIG EHGDSSVPLW SGVNVAGVAL KTLDPKLGTD SDKEHWKNIH KQVIQSAYEI IKLKGYTSWA IGLSVMDLVG SILKNLRRVH PVSTMVKGLY GIKEELFLSI PCVLGRNGVS DVVKINLNSE EEALFKKSAE TLWNIQKDLI F
Videos
SDS
Documents
Certificates
Special Offers

Special Offers