missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human LBP Protein
A cDNA sequence encoding the LBP was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP10741-10ug
Additional Details : Weight : 0.01000kg
Specifications
3929 | |
10 μg | |
< 1.0 EU per ug protein as determined by the LAL method. | |
Up to 0.2 microg/ ml LBP mediates binding of FITC-LPS (0.5microg/ml) to CD14+CHO transfectants (FACS). | |
Human | |
His | |
Recombinant Human LBP was lyophilized from a protein solution (0.3 mg/ml) containing phosphate-buffered saline, pH 7.2. |
LBP Protein | |
Research Use Only | |
LBP | |
Recombinant Protein | |
CHO | |
MGALARALPS ILLALLLTST PEALGANPGL VARITDKGLQ YAAQEGLLAL QSELLRITLP DFTGDLRIPH VGRGRYEFHS LNIHSCELLH SALRPVPGQG LSLSISDSSI RVQGRWKVRK SFFKLQGSFD VSVKGISISV NLLLGSESSG RPTVTASSCS SDIADVEVDM SGDLGWLLNL FHNQIESKFQ KVLESRICEM IQKSVSSDLQ PYLQTLPVTT EIDSFADIDY SLVEAPRATA QMLEVMFKGE IFHRNHRSPV TLLAAVMSLP EEHNKMVYFA ISDYVFNTAS LVYHEEGYLN FSITDDMIPP DSNIRLTTKS FRPFVPRLAR LYPNMNLELQ GSVPSAPLLN FSPGNLSVDP YMEIDAFVLL PSSSKEPVFR LSVATNVSAT LTFNTSKITG FLKPGKVKVE LKESKVGLFN AELLEALLNY YILNTFYPKF NDKLAEGFPL PLLKRVQLYD LGLQIHKDFL FLGANVQYMR V |