missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human LDLRAP1 Protein

A cDNA sequence encoding the LDLRAP1 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP12551-20ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15930789

  • 203.00 € / 20µg
Estimated Shipment: 24-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

LDLRAP1 Protein
Research Use Only
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MDALKSAGRA LIRSPSLAKQ SWGGGGRHRK LPENWTDTRE TLLEGMLFSL KYLGMTLVEQ PKGEELSAAA IKRIVATAKA SGKKLQKVTL KVSPRGIILT DNLTNQLIEN VSIYRISYCT ADKMHDKVFA YIAQSQHNQS LECHAFLCTK RKMAQAVTLT VAQAFKVAFE FWQVSKEEKE KRDKASQEGG DVLGARQDCT PPLKSLVATG NLLDLEETAK APLSTVSANT TNMDEVPRPQ ALSGSSVVWE LDDGLDEAFS RLAQSRTNPQ VLDTGLTAQD MHYAQCLSPV DWDKPDSSGT EQDDLFSF
Greater than 90% as determined by SDS-PAGE.
20 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
The LDLRAP1 solution (0.5 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 200mM NaCl, 2mM DTT and 10% glycerol.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers