missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human LGALS16 Protein
A cDNA sequence encoding the LGALS16 was constructed and used to recombinantly synthesize the protein.
131.00 € - 1740.00 €
Specifications
Name | LGALS16 Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Product Type | Recombinant Protein |
Cross Reactivity | Human |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15920819
|
enQuireBio™
QP12561-10UG |
10 μg |
131.00 €
10µg |
Estimated Shipment: 26-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15940819
|
enQuireBio™
QP12561-50UG |
50 μg |
203.00 €
50µg |
Estimated Shipment: 26-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15930819
|
enQuireBio™
QP12561-1MG |
1 mg |
1740.00 €
1mg |
Estimated Shipment: 26-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
LGALS16 Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
Untagged | |
1x PBS, pH 7.4. |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MSFLTVPYKL PVSLSVGSCV IIKGTLIDSS INEPQLQVDF YTEMNEDSEI AFHLRVHLGR RVVMNSREFG IWMLEENLHY VPFEDGKPFD LRIYVCLNEY EVKVNGEYIY AFVHRIPPSY VKMIQVWRDV SLDSVLVNNG RR | |
Greater than 95.0% as determined by SDS-PAGE. |