missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human LIM domain-binding protein 1

A DNA sequence encoding the Homo sapiens (Human) LIM domain-binding protein 3, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6290-ec-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16032220

  • 735.00 € / 100µg
Estimated Shipment: 27-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_001073583.1
LIM domain-binding protein 1
Research Use Only
LDB3
Recombinant Protein
E. coli
MSYSVTLTGP GPWGFRLQGG KDFNMPLTIS RITPGSKAAQ SQLSQGDLVV AIDGVNTDTM THLEAQNKIK SASYNLSLTL QKSKRPIPIS TTAPPVQTPL PVIPHQKVVV NSPANADYQE RFNPSALKDS ALSTHKPIEV KGLGGKATII HAQYNTPISM YSQDAIMDAI AGQAQAQGSD FSGSLPIKDL AVDSASPVYQ AVIKSQNKPE DEADEWARRS SNLQSRSFRI LAQMTGTEFM QDPDEEALRR SRERFETERN SPRFAKLRNW HHGLSAQILN VKS
Greater than 80% as determined by SDS-PAGE.
11155
100 μg
Not determined.
Full Length of Isoform 6
Human
GST
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers