missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human LIM domain transcription factor LMO4 Protein

A DNA sequence encoding the Homo sapiens (Human) LIM domain transcription factor LMO4, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6313-ec-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16052400

  • 3260.00 € / 1mg
Estimated Shipment: 25-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_006760.1
LIM domain transcription factor LMO4 Protein
Research Use Only
LMO4
Recombinant Protein
E. coli
MVNPGSSSQP PPVTAGSLSW KRCAGCGGKI ADRFLLYAMD SYWHSRCLKC SCCQAQLGDI GTSCYTKSGM ILCRNDYIRL FGNSGACSAC GQSIPASELV MRAQGNVYHL KCFTCSTCRN RLVPGDRFHY INGSLFCEHD RPTALINGHL NSLQSNPLLP DQKVC
Greater than 80% as determined by SDS-PAGE.
8543
1 mg
Not determined.
Full Length
Human
GST
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers