missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human LIN28 Protein
A cDNA sequence encoding the LIN28 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP12571-5ug
Additional Details : Weight : 0.01000kg
Spécification
LIN28 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
GPSVSNQQFA GGCAKAAEEA PEEAPEDAAR AADEPQLLHG AGICKWFNVR MGFGFLSMTA RAGVALDPPV DVFVHQSKLH MEGFRSLKEG EAVEFTFKKS AKGLESIRVT GPGGVFCIGS ERRPKGKSMQ KRRSKGDRCY NCGGLDHHAK ECKLPPQPKK CHFCQSISHM VASCPLKAQQ GPSAQGKPTY FREEEEEIHS PTLLPEAQNG GYGRKKRRQR RR | |
Greater than 90% as determined by SDS-PAGE (coomassie staining). |
5 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
Untagged | |
The LIN28 protein solution was formulated in PBS with 50mM arginine |