missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Human Lipoprotein Lipase/LPL Protein
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
Description
The Recombinant Human Lipoprotein Lipase/LPL Protein is derived from CHO. The Recombinant Human Lipoprotein Lipase/LPL Protein has been validated for the following applications: Enzyme Activity.
Specifications
Specifications
| Accession Number | P06858 |
| Gene ID (Entrez) | 4023 |
| Molecular Weight (g/mol) | 51 kDa |
| Name | EC 3.1.1, HDLCQ11, LIPD, LIPDEC 3.1.1.34, lipoprotein lipase, LPL |
| Quantity | 100 μg |
| Source | Chinese Hamster Ovary cell line,CHO-derived human Lipoprotein Lipase/LPL protein Ala28-Gly475 with a C-terminal 6-His tag |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -70°C as supplied. 3 months, -70°C under sterile conditions after opening. |
| Endotoxin Concentration | <0.10 EU per 1 µg of the protein by the LAL method. |
| Conjugate | Unconjugated |
| Purity or Quality Grade | >95%, by SDS-PAGE under reducing conditions and visualized by silver stain. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction