missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Lipoyl synthase, mitochondrial Protein

A DNA sequence encoding the Homo sapiens (Human) Lipoyl synthase, mitochondrial, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6307-ec-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16032360

  • 2285.00 € / 1mg
Estimated Shipment: 21-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_001265519.1
Lipoyl synthase, mitochondrial Protein
Research Use Only
LIAS
Recombinant Protein
E. coli
LSSLPDKKKE LLQNGPDLQD FVSGDLADRS TWDEYKGNLK RQKGERLRLP PWLKTEIPMG KNYNKLKNTL RNLNLHTVCE EARCPNIGEC WGGGEYATAT ATIMLMGDTC TRGCRFCSVK TARNPPPLDA SEPYNTAKAI AEWGLDYVVL TSVDRDDMPD GGAEHIAKTV SYLKERNPKI LVECLTPDFR GDLKAIEKVA LSGLDVYAHN VETVPELQSK VRDPRANFDQ SLRVLKHAKK VQPDVISKTS IMLGLGENDE QVYATMKALR EADVDCLTLG QYMQPTRRHL KVEEYITPEK FKYWEKVGNE LGFHYTASGP LVRSSYKAGE FFLKNLVAKR KTKDL
Greater than 90% as determined by SDS-PAGE.
11019
1 mg
Not determined.
Full Length
Human
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers