missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human LPCAT1 Protein
A cDNA sequence encoding the LPCAT1 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP12580-2ug
Additional Details : Weight : 0.01000kg
Specifications
79888 | |
2 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSSAEKEPE QPPALWRKVV DFLLKAIMRT MWFAGGFHRV AVKGRQALPT EAAILTLAPH SSYFDAIPVT MTMSSIVMKA ESRDIPIWGT LIQYIRPVFV SRSDQDSRRK TVEEIKRRAQ SNGKWPQIMI FPEGTCTNRT CLITFKPGAF IPGAPVQPVV LRYPNKLDTI TWTWQGPGAL EILWLTLCQF HNQVEIEFLP VYSPSEEEKR NPALYASNVR RVMAEALGVS VTDYTFEDCQ LALAEGQLRL PADTCLLEFA RLVRGLGLKP EKLEKDLDRY SERARMKGGE KIGIAEFAAS LEVPVSDLLE DMFSLFDESG SGEVDLRECV VALSVVCRPA RTLDTIQLAF KMYGAQEDGS VGEGDLSCIL KTALGVAELT VTDLFRAIDQ EEKGKITFAD FHRFAEMYPA FAEEYLYPDQ THFESCAETS PAPIPNGFCA DFSPENSDAG RKPVRKKLD | |
Greater than 90.0% as determined by SDS-PAGE. |
LPCAT1 Protein | |
Research Use Only | |
LPCAT1 | |
Human | |
His | |
The LPCAT1 solution (0.25 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 1M Urea and 10% glycerol. |