missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human LRRC59 Protein

A cDNA sequence encoding the LRRC59 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP12587-2ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15960909

  • 131.00 € / 2µg
Estimated Shipment: 26-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

LRRC59 Protein
Research Use Only
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MGSMTKAGSK GGNLRDKLDG NELDLSLSDL NEVPVKELAA LPKATILDLS CNKLTTLPSD FCGLTHLVKL DLSKNKLQQL PADFGRLVNL QHLDLLNNKL VTLPVSFAQL KNLKWLDLKD NPLDPVLAKV AGDCLDEKQC KQCANKVLQH MKAVQADQER ERQRRLEVER EAEKKREAKQ RAKEAQEREL RKREKAEEKE RRRKEYDALK AAKREQEKKP KKEANQAPKS KSGSRPRKPP PRKHTRS
Greater than 90.0% as determined by SDS-PAGE.
2 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
The LRRC59 solution (0.25 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 0.2M NaCl, 2mM DTT, 2mM EDTA and 50% glycerol.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers