missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Lutropin-choriogonadotropic hormone receptor Protein

A DNA sequence encoding the Homo sapiens (Human) Lutropin-choriogonadotropic hormone receptor, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6305-ec-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16042330

  • 505.00 € / 100µg
Estimated Shipment: 21-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_000224.2
Lutropin-choriogonadotropic hormone receptor Protein
Research Use Only
Lhcgr
Recombinant Protein
E. coli
EALCPEPCNC VPDGALRCPG PTAGLTRLSL AYLPVKVIPS QAFRGLNEVI KIEISQIDSL ERIEANAFDN LLNLSEILIQ NTKNLRYIEP GAFINLPRLK YLSICNTGIR KFPDVTKVFS SESNFILEIC DNLHITTIPG NAFQGMNNES VTLKLYGNGF EEVQSHAFNG TTLTSLELKE NVHLEKMHNG AFRGATGPKT LDISSTKLQA LPSYGLESIQ RLIATSSYSL KKLPSRETFV NLLEATLTYP SHCCAFRNLP TKEQNFSHSI SENFSKQCES TVRKVNNKTL YSSMLAESEL SGWDYEYGFC LPKTPRCAPE PDAFNPCEDI MGYDFLR
Greater than 90% as determined by SDS-PAGE.
3973
100 μg
Not determined.
Extracellular Domain
Human
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers