missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human LY86 Protein
A cDNA sequence encoding the LY86 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP12602-5ug
Additional Details : Weight : 0.01000kg
Specifications
LY86 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSGGGGKAW PTHVVCSDSG LEVLYQSCDP LQDFGFSVEK CSKQLKSNIN IRFGIILRED IKELFLDLAL MSQGSSVLNF SYPICEAALP KFSFCGRRKG EQIYYAGPVN NPEFTIPQGE YQVLLELYTE KRSTVACANA TIMCS | |
Greater than 85% as determined by SDS-PAGE. |
5 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The LY86 solution (1 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 0.4M Urea and 10% glycerol. |