missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human LYN Protein
A cDNA sequence encoding the LYN was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP12605-5ug
Additional Details : Weight : 0.01000kg
Specifications
LYN Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMGCIKSK GKDSLSDDGV DLKTQPVRNT ERTIYVRDPT SNKQQRPVPE SQLLPGQRFQ TKDPEEQGDI VVALYPYDGI HPDDLSFKKG EKMKVLEEHG EWWKAKSLLT KKEGFIPSNY VAKLNTLETE EWFFKDITRK DAERQLLAPG NSAGAFLIRE SETLKGSFSL SVRDFDPVHG DVIKHYKIRS LDNGGYYISP RITFPCISDM IKHYQKQADG LCRRLEKACI SPKPQKPWDK DAWEIPRESI KLVKRLGAGQ FGEVWMGYYN NSTKVAVKTL KPGTMSVQAF LEEANLMKTL QHDKLVRLYA VVTREEPIYI ITEYMAKGSL LDFLKSDEGG KVLLPKLIDF SAQIAEGMAY IERKNYIHRD LRAANVLVSE SLMCKIADFG LARVIEDNEY TAREGAKFPI KWTAPEAINF GCFTIKSDVW SFGILLYEIV TYGKIPYPGR TNADVMTALS QGYRMPRVEN CPDELYDIMK MCWKEKAEER PTFDYLQSVL DDFYTATEGQ YQQQP | |
Greater than 90.0% as determined by SDS-PAGE. |
5 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The LYN solution (1 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0) ,0.4M urea and 10% glycerol. |