missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Lysine-specific demethylase 5A Protein

A DNA sequence encoding the Homo sapiens (Human) Lysine-specific demethylase 5A, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6257-ye-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16031980

  • 615.00 € / 100µg
Estimated Shipment: 24-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_001036068.1
Lysine-specific demethylase 5A Protein
Research Use Only
KDM5A
Recombinant Protein
Yeast
EYALSGWNLN NMPVLEQSVL AHINVDISGM KVPWLYVGMC FSSFCWHIED HWSYSINYLH WGEPKTWYGV PSHAAEQLEE VMRELAPELF ESQPDLLHQL VTIMNPNVLM EHGVPVYRTN QCAGEFVVTF PRAYHSGFNQ GYNFAEAVNF CTADWLPIGR QCVNHYR
Greater than 90% as determined by SDS-PAGE.
5927
100 μg
Not determined.
Full Length
Human
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers