missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Lysyl oxidase homolog 4 Protein

A DNA sequence encoding the Homo sapiens (Human) Lysyl oxidase homolog 4, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7943-ec-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16031751

  • 3375.00 € / 1mg
Estimated Shipment: 28-07-2022
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_115587.6
Lysyl oxidase homolog 4 Protein
1 mg
Not determined.
Full Length
Human
His
Tris-based buffer, 50% glycerol
84171
Greater than 90% as determined by SDS-PAGE.
Research Use Only
Loxl4
Recombinant Protein
E. coli
QSLGTTKLRL VGPESKPEEG RLEVLHQGQW GTVCDDNFAI QEATVACRQL GFEAALTWAH SAKYGQGEGP IWLDNVRCVG TESSLDQCGS NGWGVSDCSH SEDVGVICHP RRHRGYLSET VSNALGPQGR RLEEVRLKPI LASAKQHSPV TEGAVEVKYE GHWRQVCDQG WTMNNSRVVC GMLGFPSEVP VDSHYYRKVW DLKMRDPKSR LKSLTNKNSF WIHQVTCLGT EPHMANCQVQ VAPARGKLRP ACPGGMHAVV SCVAGPHFRP PKTKPQRKGS WAEEPRVRLR SGAQVGEGRV EVLMNRQWGT VCDHRWNLIS ASVVCRQLGF GSAREALFGA RLGQGLGPIH LSEVRCRGYE RTLSDCPALE GSQNGCQHEN DAAVRCNVPN MGFQNQVRLA GGRIPEEGLL EVQVEVNGVP RWGSVCSENW GLTEAMVACR QLGLGFAIHA YKETWFWSGT PRAQEVVMSG VRCSGTELAL QQCQRHGPVH CSHGGGRFLA GVSCMDSAPD LVMNAQLVQE TAYLEDRPLS QLYCAHEENC LSKSADHMDW PYGYRRLLRF STQIYNLGRT DFRPKTGRDS WVWHQCHRHY HSIEVFTHYD LLTLNGSKVA EGHKASFCLE DTNCPTGLQR RYACANFGEQ GVTVGCWDTY RHDIDCQWVD ITDVGPGNYI FQVIVNPHYE VAESDFSNNM LQCRCKYDGH RVWLHNCHTG NSYPANAELS LEQEQRLRNN LI
Videos
SDS
Documents
Certificates
Special Offers

Special Offers