missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Human M-CSF, Animal-Free Protein

Product Code. 18777196
Change view
Click to view available options
Quantity:
25 μg
5 μg
500 μg
Unit Size:
25µg
500µg
5µg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18777196 500 μg 500µg
18787616 5 μg 5µg
18748446 25 μg 25µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 18777196 Supplier R&D Systems™ Supplier No. AFL216500
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Measured in a cell proliferation assay using M-NFS-60 mouse myelogenous leukemia lymphoblast cells

  • ED50 for this effect is 0.5 to 1.5 ng/mL.
  • The specific activity of Recombinant Human M-CSF is approximately 1.68 x 105 IU/μg, which is calibrated against recombinant human M-CSF WHO International Standard (NIBSC code: 89/512).
TRUSTED_SUSTAINABILITY

Specifications

Accession Number NP_757350.2
For Use With (Application) Bioactivity
Gene ID (Entrez) 1435
Molecular Weight (g/mol) 18.5 kDa (monomer)
Name colony stimulating factor 1 (macrophage), CSF1, CSF-1, Lanimostim, macrophage colony stimulating factor, macrophage colony-stimulating factor 1, MCSF, M-CSF, MCSFlanimostim, MGC31930
Quantity 500 μg
Source E. coli-derived human M-CSF protein Glu33-Ser190, with an N-terminal Met Produced using non-animal reagents in an animal-free laboratory.
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 3 months, -20 to -70 °C under sterile conditions after reconstitution.
Gene Symbol CSF1
Conjugate Unconjugated
Species Human
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Buffer Lyophilized from a 0.2 μm filtered solution in PBS.
Endotoxin Level <0.01 EU per 1 μg of the protein by the LAL method.
Product Line R&D Systems
Purity or Quality Grade >97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.