missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Human M-CSF, Animal-Free Protein

Product Code. p-7182944
Click to view available options
Quantity:
25 μg
5 μg
500 μg
Unit Size:
25µg
500µg
5µg
This item is not returnable. View return policy

Product Code. 18787616

Brand: R&D Systems™ AFL216005

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Measured in a cell proliferation assay using M-NFS-60 mouse myelogenous leukemia lymphoblast cells

  • ED50 for this effect is 0.5 to 1.5 ng/mL.
  • The specific activity of Recombinant Human M-CSF is approximately 1.68 x 105 IU/μg, which is calibrated against recombinant human M-CSF WHO International Standard (NIBSC code: 89/512).
TRUSTED_SUSTAINABILITY

Specifications

Accession Number NP_757350.2
For Use With (Application) Bioactivity
Gene ID (Entrez) 1435
Molecular Weight (g/mol) 18.5 kDa (monomer)
Name colony stimulating factor 1 (macrophage), CSF1, CSF-1, Lanimostim, macrophage colony stimulating factor, macrophage colony-stimulating factor 1, MCSF, M-CSF, MCSFlanimostim, MGC31930
Quantity 5 μg
Source E. coli-derived human M-CSF protein Glu33-Ser190, with an N-terminal Met Produced using non-animal reagents in an animal-free laboratory.
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 3 months, -20 to -70 °C under sterile conditions after reconstitution.
Gene Symbol CSF1
Conjugate Unconjugated
Species Human
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Buffer Lyophilized from a 0.2 μm filtered solution in PBS.
Endotoxin Level <0.01 EU per 1 μg of the protein by the LAL method.
Purity or Quality Grade >97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.