missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Human M-CSF Protein

Product Code. 18738636
Change view
Click to view available options
Quantity:
10 μg
100 μg
Unit Size:
100µg
10µg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18738636 10 μg 10µg
18706546 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18738636 Supplier R&D Systems™ Supplier No. 216MC010/CF

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

Measured in a cell proliferation assay using M-NFS-60 mouse myelogenous leukemia lymphoblast cells. Nakoinz, I. et al. (1990) J. Immunol. 145:860. The ED50 for this effect is 0.5-1.5 ng/mL.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number NP_757350
For Use With (Application) Bioactivity
Formulation Lyophilized from a 0.2 μm filtered solution in PBS.
Gene ID (Entrez) 1435
Molecular Weight (g/mol) 18.5 kDa (monomer)
Quantity 10 μg
Source <EM>E. coli</EM>-derived human M-CSF protein Glu33-Ser190, with an N-terminal Met
Storage Requirements Manual defrost freezer; Avoid repeated freeze-thaw cycles; 12 mo. from receipt, -20 to -70°C as supplied; 1 mo, 2 to 8°C under sterile conditions after reconstitution; 3 mo, -20 to -70°C under sterile conditions after reconstitution
Regulatory Status RUO
Endotoxin Concentration <1.0 EU per 1 μg of the protein by the LAL method.
Gene Symbol CSF1
Conjugate Unconjugated
Cross Reactivity M-CSF
Species Human
Reconstitution Reconstitute at 50-500 μg/mL in sterile PBS.
Purity or Quality Grade >97%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.