missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human MAGE-1 Protein

Product Code. 16057941
Change view
Click to view available options
Quantity:
100 μg
20 μg
50 μg
Unit Size:
100µg
20µg
50µg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16057941 20 μg 20µg
16067941 50 μg 50µg
16047941 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16057941 Supplier enQuireBio™ Supplier No. QP9261ma20ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

A DNA sequence encoding the Homo sapiens (Human) MAGE-1, was expressed in the hosts and tags indicated.

Specifications

Accession Number NP_004979.3
Gene ID (Entrez) 4100
Name MAGE-1 Protein
Quantity 20 μg
Regulatory Status Research Use Only
Endotoxin Concentration Not determined.
Gene Symbol MAGEA1
Protein Length Full Length
Product Type Recombinant Protein
Cross Reactivity Human
Species Mammal
Protein Tag Flag
Sequence MSLEQRSLHC KPEEALEAQQ EALGLVCVQA ATSSSSPLVL GTLEEVPTAG STDPPQSPQG ASAFPTTINF TRQRQPSEGS SSREEEGPST SCILESLFRA VITKKVADLV GFLLLKYRAR EPVTKAEMLE SVIKNYKHCF PEIFGKASES LQLVFGIDVK EADPTGHSYV LVTCLGLSYD GLLGDNQIMP KTGFLIIVLV MIAMEGGHAP EEEIWEELSV MEVYDGREHS AYGEPRKLLT QDLVQEKYLE YRQVPDSDPA RYEFLWGPRA LAETSYVKVL EYVIKVSARV RFFFPSLREA ALREEEEGV
Buffer Tris-based buffer, 50% glycerol
Purity or Quality Grade Greater than 90% as determined by SDS-PAGE.
Mostrar más Mostrar menos
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.