missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human MAGE-1 Protein
A DNA sequence encoding the Homo sapiens (Human) MAGE-1, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP9261-ma-100ug
Additional Details : Weight : 0.01000kg
Specifications
NP_004979.3 | |
MAGE-1 Protein | |
Research Use Only | |
MAGEA1 | |
Recombinant Protein | |
Human | |
MSLEQRSLHC KPEEALEAQQ EALGLVCVQA ATSSSSPLVL GTLEEVPTAG STDPPQSPQG ASAFPTTINF TRQRQPSEGS SSREEEGPST SCILESLFRA VITKKVADLV GFLLLKYRAR EPVTKAEMLE SVIKNYKHCF PEIFGKASES LQLVFGIDVK EADPTGHSYV LVTCLGLSYD GLLGDNQIMP KTGFLIIVLV MIAMEGGHAP EEEIWEELSV MEVYDGREHS AYGEPRKLLT QDLVQEKYLE YRQVPDSDPA RYEFLWGPRA LAETSYVKVL EYVIKVSARV RFFFPSLREA ALREEEEGV | |
Greater than 90% as determined by SDS-PAGE. |
4100 | |
100 μg | |
Not determined. | |
Full Length | |
Human | |
Flag | |
Tris-based buffer, 50% glycerol |