missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human MAGED1 Protein

A cDNA sequence encoding the MAGED1 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP12628-1ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15961039

  • 131.00 € / 1µg
Estimated Shipment: 25-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

MAGED1 Protein
Research Use Only
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MGSLRPSPNS RASQNPGAAQ PRDVALLQER ANKLVKYLML KDYTKVPIKR SEMLRDIIRE YTDVYPEIIE RACFVLEKKF GIQLKEIDKE EHLYILISTP ESLAGILGTT KDTPKLGLLL VILGVIFMNG NRASEAVLWE ALRKMGLRPG VRHPLLGDLR KLLTYEFVKQ KYLDYRRVPN SNPPEYEFLW GLRSYHETSK MKVLRFIAEV QKRDPRDWTA QFMEAADEAL DALDAAAAEA EARAEARTRM GIGDEAVSGP
Greater than 85% as determined by SDS-PAGE.
1 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
MAGED1 protein solution (0.25 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 20% glycerol and 1mM DTT.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers