missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human MAP1LC3A Protein

A cDNA sequence encoding the MAP1LC3A was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP12634-5ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15961059

  • 131.00 € / 5µg
Estimated Shipment: 05-07-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

MAP1LC3A Protein
Research Use Only
Recombinant Protein
E. coli
MPSDRPFKQR RSFADRCKEV QQIRDQHPSK IPVIIERYKG EKQLPVLDKT KFLVPDHVNM SELVKIIRRR LQLNPTQAFF LLVNQHSMVS VSTPIADIYE QEKDEDGFLY MVYASQETFG LEHHHHHH
Greater than 95.0% as determined by SDS-PAGE.
5 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
The MAP1LC3A solution (1 mg/ml) contains 20mM Tris-HCl pH 8.0, 1mM DTT, 0.1M NaCl and 20% glycerol.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers