missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human MAP2K2 Protein
Click to view available options
Quantity:
1 μg
5 μg
50 μg
Unit Size:
1µg
50µg
5µg
Specifications
Specifications
| Name | MAP2K2 Protein |
| Quantity | 1 μg |
| Regulatory Status | Research Use Only |
| Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
| Product Type | Recombinant Protein |
| Cross Reactivity | Human |
| Species | E. coli |
| Protein Tag | His |
| Sequence | MGSSHHHHHH SSGLVPRGSH MGSHMLARRK PVLPALTINP TIAEGPSPTS EGASEANLVD LQKKLEELEL DEQQKKRLEA FLTQKAKVGE LKDDDFERIS ELGAGNGGVV TKVQHRPSGL IMARKLIHLE IKPAIRNQII RELQVLHECN SPYIVGFYGA FYSDGEISIC MEHMDGGSLD QVLKEAKRIP EEILGKVSIA VLRGLAYLRE KHQIMHRDVK PSNILVNSRG EIKLCDFGVS GQLIDSMANS FVGTRSYMAP ERLQGTHYSV QSDIWSMGLS LVELAVGRYP IPPPDAKELE AIFGRPVVDG EEGEPHSISP RPRPPGRPVS GHGMDSRPAM AIFELLDYIV NEPPPKLPNG VFTPDFQEFV NKCLIKNPAE RADLKMLTNH TFIKRSEVEE VDFAGWLCKT LRLNQPGTPT RTAV |
| Buffer | MAP2K2 protein solution (0.25 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 0.1M NaCl and 10% glycerol. |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction