missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human MAT2B Protein

A cDNA sequence encoding the MAT2B was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP12651-20ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15991109

  • 203.00 € / 20µg
Estimated Shipment: 07-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

MAT2B Protein
Research Use Only
Recombinant Protein
E. coli
MPEMPEDMEQ EEVNIPNRRV LVTGATGLLG RAVHKEFQQN NWHAVGCGFR RARPKFEQVN LLDSNAVHHI IHDFQPHVIV HCAAERRPDV VENQPDAASQ LNVDASGNLA KEAAAVGAFL IYISSDYVFD GTNPPYREED IPAPLNLYGK TKLDGEKAVL ENNLGAAVLR IPILYGEVEK LEESAVTVMF DKVQFSNKSA NMDHWQQRFP THVKDVATVC RQLAEKRMLD PSIKGTFHWS GNEQMTKYEM ACAIADAFNL PSSHLRPITD SPVLGAQRPR NTQLDCSKLE TLGIGQRTPF RIGIKESLWP FLIDKRWRQT VFH
Greater than 90.0% as determined by SDS-PAGE.
20 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
Untagged
The MAT2B protein solution contains 20mM Tris-HCl, pH 8.0, 1mM DTT, 1mM EDTA and 10% Glycerol.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers