missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human MCEE Protein
A cDNA sequence encoding the MCEE was constructed and used to recombinantly synthesize the protein.
131.00 € - 3915.00 €
Specifications
Gene ID (Entrez) | 84693 |
---|---|
Name | MCEE Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Gene Symbol | MCEE |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Código de producto | Marca | Quantity | Precio | Cantidad y disponibilidad | |||||
15981139
|
enQuireBio™
QP12659-5UG |
5 μg |
131.00 €
5µg |
Estimated Shipment: 20-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15971139
|
enQuireBio™
QP12659-20UG |
20 μg |
203.00 €
20µg |
Estimated Shipment: 20-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15961139
|
enQuireBio™
QP12659-1MG |
1 mg |
3915.00 €
1mg |
Estimated Shipment: 20-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Especificaciones
84693 | |
Research Use Only | |
MCEE | |
Human | |
His | |
The MCEE protein solution (1 mg/1ml) is formulated in 20mM Tris-HCl buffer (pH 8.0), 0.2M NaCl, 1mM DTT, 0.1mM PMSF and 10% glycerol. |
MCEE Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MQVTGSVWNL GRLNHVAIAV PDLEKAAAFY KNILGAQVSE AVPLPEHGVS VVFVNLGNTK MELLHPLGRD SPIAGFLQKN KAGGMHHICI EVDNINAAVM DLKKKKIRSL SEEVKIGAHG KPVIFLHPKD CGGVLVELEQ A | |
Greater than 90% as determined by SDS-PAGE. |