missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human MECR Protein
Click to view available options
Quantity:
1 mg
10 μg
2 μg
Unit Size:
10µg
1mg
2µg
Specifications
Specifications
| Name | MECR Protein |
| Quantity | 10 μg |
| Regulatory Status | Research Use Only |
| Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
| Product Type | Recombinant Protein |
| Cross Reactivity | Human |
| Species | E. coli |
| Protein Tag | His |
| Sequence | MGSSHHHHHH SSGLVPRGSH MPAKVVELKN LELAAVRGSD VRVKMLAAPI NPSDINMIQG NYGLLPELPA VGGNEGVAQV VAVGSNVTGL KPGDWVIPAN AGLGTWRTEA VFSEEALIQV PSDIPLQSAA TLGVNPCTAY RMLMDFEQLQ PGDSVIQNAS NSGVGQAVIQ IAAALGLRTI NVVRDRPDIQ KLSDRLKSLG AEHVITEEEL RRPEMKNFFK DMPQPRLALN CVGGKSSTEL LRQLARGGTM VTYGGMAKQP VVASVSLLIF KDLKLRGFWL SQWKKDHSPD QFKELILTLC DLIRRGQLTA PACSQVPLQD YQSALEASMK PFISSKQILT M |
| Buffer | MECR Human solution containing 20mM Trsi pH 8.0, 0.2M NaCl, 5mM DTT and 20% glycerol. |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction