missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human MET Protein

Product Code. 15931219
Click to view available options
Quantity:
10 μg
2 μg
5 μg
Unit Size:
10µg
2µg
5µg
This item is not returnable. View return policy

Product Code. 15931219

Brand: enQuireBio™ QP12683GST5ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

A cDNA sequence encoding the MET was constructed and used to recombinantly synthesize the protein.

Specifications

Gene ID (Entrez) 4233
Name MET Protein
Quantity 5 μg
Regulatory Status Research Use Only
Endotoxin Concentration Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Gene Symbol MET
Product Type Recombinant Protein
Cross Reactivity Human
Species E. coli
Protein Tag GST
Sequence MKAPAVLAPG ILVLLFTLVQ RSNGECKEAL AKSEMNVNMK YQLPNFTAET PIQNVILHEH HIFLGATNYI YVLNEEDLQK VAEYKTGPVL EHPDCFPCQD CSSKANLSGG VWKDNINMAL VVDTYYDDQL ISCGSVNRGT CQRHVFPHNH TADIQSEVHC IFSPQIEEPS QCPDCVVSAL GAKVLSSVKD RFINFFVGNT INSSYFPDHP LHSISVRRLK ETKDGFMFLT DQSYIDVLPE FRDSYPIKYV HAFESNNFIY FLTVQRETLD AQTFHTRIIR FCSINSGLHS YMEMPLECIL TEKRKKRSTK KEVFNILQAA YVSKPGAQLA RQIGASLNDD ILFGVFAQSK PDSAEPMDRS AMCAFPIKYV NDFFNKIVNK NNVRCLQHFY GPNHEHCFNR TLLRNSSGCE ARRDEYRTEF TTALQRVDLF MGQFSEVLLT SISTFIKGDL TIANLGTSEG RFMQVVVSRS GPSTPHVNFL LDSHPVSPEV IVEHTLNQNG YTLVITGKKI TKIPLNGLGC RHFQSCSQCL SAPPFVQCGW CHDKCVRSEE CLSGTWTQQI CLPAIYKVFP NSAPLEGGTR LTICGWDFGF RRNNKFDLKK TRVLLGNESC TLTLSESTMN TLKCTVGPAM NKHFNMSIII SNGHGTTQYS TFSYVDPVIT SISPKYGPMA GGTLLTLTGN YLNSGNSRHI SIGGKTCTLK SVSNSILECY TPAQTISTEF AVKLKIDLAN RETSIFSYRE DPIVYEIHPT KSFISGGSTI TGVGKNLNSV SVPRMVINVH EAGRNFTVAC QHRSNSEIIC CTTPSLQQLN LQLPLKTKAF FMLDGILSKY FDLIYVHNPV FKPFEKPVMI SMGNENVLEI KGNDIDPEAV KGEVLKVGNK SCENIHLHSE AVLCTVPNDL LKLNSELNIE WKQAISSTVL GKVIVQPDQN FTGLIAGVVS ISTALLLLLG FFLWLKKRKQ IKDLGSELVR YDARVHTPHL DRLVSARSVS PTTEMVSNES VDYRATFPED QFPNSSQNGS CRQVQYPLTD MSPILTSGDS DISSPLLQNT VHIDLSALNP ELVQAVQHVV IGPSSLIVHF NEVIGRGHFG CVYHGTLLDN DGKKIHCAVK SLNRITDIGE VSQFLTEGII MKDFSHPNVL SLLGICLRSE GSPLVVLPYM KHGDLRNFIR NETHNPTVKD LIGFGLQVAK GMKYLASKKF VHRDLAARNC MLDEKFTVKV ADFGLARDMY DKEYYSVHNK TGAKLPVKWM ALESLQTQKF TTKSDVWSFG VLLWELMTRG APPYPDVNTF DITVYLLQGR RLLQPEYCPD PLYEVMLKCW HPKAEMRPSF SELVSRISAI FSTFIGEHYV HVNATYVNVK CVAPYPSLLS SEDNADDEVD TRPASFWETS
Buffer MET is supplied in 100microg/ml in 50mM Tris-HCl (pH 7.5) with 10mM L-glutathione (reduced).
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.