missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Methylthioribulose-1-phosphate dehydratase Protein
A DNA sequence encoding the Homo sapiens (Human) Methylthioribulose-1-phosphate dehydratase, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7896-ec-1mg
Additional Details : Weight : 0.01000kg
Specifications
NP_057041.2 | |
Methylthioribulose-1-phosphate dehydratase Protein | |
Research Use Only | |
APIP | |
Recombinant Protein | |
E. coli | |
MSGCDAREGD CCSRRCGAQD KEHPRYLIPE LCKQFYHLGW VTGTGGGISL KHGDEIYIAP SGVQKERIQP EDMFVCDINE KDISGPSPSK KLKKSQCTPL FMNAYTMRGA GAVIHTHSKA AVMATLLFPG REFKITHQEM IKGIKKCTSG GYYRYDDMLV VPIIENTPEE KDLKDRMAHA MNEYPDSCAV LVRRHGVYVW GETWEKAKTM CECYDYLFDI AVSMKKVGLD PSQLPVGENG IV | |
Greater than 80% as determined by SDS-PAGE. |
51074 | |
1 mg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |