missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human METTL1 Protein

A cDNA sequence encoding the METTL1 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP12687-10ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15931229

  • 181.00 € / 10µg
Estimated Shipment: 23-01-2023
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

METTL1 Protein
10 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
The METTL1 protein solution (0.5 mg/1ml) is formulated in 20mM Tris-HCl buffer (pH 8.0)1mM DTT, 50mM NaCl and 10% glycerol.
Greater than 90% as determined by SDS-PAGE.
Research Use Only
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MAAETRNVAG AEAPPPQKRY YRQRAHSNPM ADHTLRYPVK PEEMDWSELY PEFFAPLTQN QSHDDPKDKK EKRAQAQVEF ADIGCGYGGL LVELSPLFPD TLILGLEIRV KVSDYVQDRI RALRAAPAGG FQNIACLRSN AMKHLPNFFY KGQLTKMFFL FPDPHFKRTK HKWRIISPTL LAEYAYVLRV GGLVYTITDV LELHDWMCTH FEEHPLFERV PLEDLSEDPV VGHLGTSTEE GKKVLRNGGK NFPAIFRRIQ DPVLQAVTSQ TSLPGH
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers