missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human MFG-E8 / lactadherin Protein

A DNA sequence encoding the Homo sapiens (Human) MFG-E8 / lactadherin, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6356-ec-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16042710

  • 2365.00 € / 1mg
Estimated Shipment: 02-08-2022
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_001108086.1
MFG-E8 / lactadherin Protein
1 mg
Not determined.
Full Length
Human
His-SUMO
Tris-based buffer, 50% glycerol
4240
Greater than 90% as determined by SDS-PAGE.
Research Use Only
MFGE8
Recombinant Protein
E. coli
LDICSKNPCH NGGLCEEISQ EVRGDVFPSY TCTCLKGYAG NHCETKCVEP LGMENGNIAN SQIAASSVRV TFLGLQHWVP ELARLNRAGM VNAWTPSSND DNPWIQVNLL RRMWVTGVVT QGASRLASHE YLKAFKVAYS LNGHEFDFIH DVNKKHKEFV GNWNKNAVHV NLFETPVEAQ YVRLYPTSCH TACTLRFELL GCELNGCANP LGLKNNSIPD KQITASSSYK TWGLHLFSWN PSYARLDKQG NFNAWVAGSY GNDQWLQVDL GSSKEVTGII TQGARNFGSV QFVASYKVAY SNDSANWTEY QDPRTGSSKI FPGNWDNHSH KKNLFETPIL ARYVRILPVA WHNRIALRLE LLGC
Videos
SDS
Documents
Certificates
Special Offers

Special Offers