missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human MIA2 Protein
A cDNA sequence encoding the MIA2 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP12694-5ug
Additional Details : Weight : 0.01000kg
Specifications
MIA2 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MLESTKLLAD LKKCGDLECE ALINRVSAMR DYRGPDCRYL NFTKGEEISV YVKLAGERED LWAGSKGKEF GYFPRDAVQI EEVFISEEIQ MSTKESDFLC L | |
Greater than 97.0% as determined by(a) Analysis by RP-HPLC. (b) Analysis by SDS-PAGE. |
5 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
Untagged | |
The MIA2 protein was lyophilized from a concentrated (1 mg/ml) solution containing 20mM Phosphate buffer pH=7.4 and 150mM NaCl. |