missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Midkine / MDK Protein

A DNA sequence encoding the Homo sapiens (Human) Midkine / MDK Protein, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8694-ec-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16045991

  • 505.00 € / 100µg
Estimated Shipment: 10-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_001012333.1
Midkine / MDK Protein
Research Use Only
MDK
Recombinant Protein
E. coli
VAKKKDKVKK GGPGSECAEW AWGPCTPSSK DCGVGFREGT CGAQTQRIRC RVPCNWKKEF GADCKYKFEN WGACDGGTGT KVRQGTLKKA RYNAQCQETI RVTKPCTPKT KAKAKAKKGK GKD
Greater than 90% as determined by SDS-PAGE.
4192
100 μg
Not determined.
Full Length
Human
GST
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers